Assay Details
| Gene Symbol: Ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133647.1 | - | 1880 | 95 | ||
| M37890.1 | - | 3 - 4 | 1819 | 95 | ||
| S56585.1 | - | 3 - 4 | 1898 | 95 | ||
| X53779.1 | - | 3 - 4 | 1851 | 95 | ||
| X59411.1 | - | 3 - 4 | 1850 | 95 | ||
| X59592.1 | - | 3 - 4 | 1939 | 95 | ||
More Information
Set Membership: |
|

Ar
Genomic Map