Assay Details
| Gene Symbol: Chst10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145142.2 | NP_660124.2 | 5 - 6 | 846 | 111 | |
| GenBank mRNA | AF360543.1 | - | 5 - 6 | 678 | 111 | |
| AK046415.1 | - | 5 - 6 | 698 | 111 | ||
| AK050676.1 | - | 4 - 5 | 728 | 111 | ||
| BC026960.1 | - | 3 - 4 | 355 | 111 | ||
| BC056956.1 | - | 5 - 6 | 775 | 111 | ||
| BC132223.1 | - | 4 - 5 | 518 | 40130500 | 111 | |
| BC132225.1 | - | 4 - 5 | 518 | 40130502 | 111 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chst10
Genomic Map