Assay Details
| Gene Symbol: 4933427E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045853.1 | 4 - 5 | 746 | 114 | ||
| GenBank mRNA | AK016948.1 | - | 4 - 5 | 114 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 12069 | Gm12069 | OTTMUSG00000005297 | -34.7kb | Sense | Chr. 11 - 25301259 - 25322784 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 5730522E02 gene | 5730522E02Rik | 258.6kb | Anti-sense | Chr. 11 - 25616846 - 26210576 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933427E13Rik
Genomic Map