Assay Details
| Gene Symbol: 4933427E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045853.1 | 6 - 7 | 1025 | 107 | ||
| GenBank mRNA | AK016948.1 | - | 6 - 7 | 1026 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 12069 | Gm12069 | OTTMUSG00000005297 | -42.5kb | Sense | Chr. 11 - 25301259 - 25322784 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 5730522E02 gene | 5730522E02Rik | 250.5kb | Anti-sense | Chr. 11 - 25616846 - 26210576 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152001 | AK016948.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423904 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760359 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260392 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105420 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118665 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152001 | AK016948.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07282905 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02282077 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105420 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118665 | - | - |
| Whole Mouse Genome G4122A | A_52_P533889 | AK016948.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104351 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105420 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09075958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09341581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09622990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09295436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09274874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09621977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09370936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09622981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09476048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09558713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09207784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09370096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09622992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09508494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09303337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09370946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09036333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09508478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09295435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27656509 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1431568_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431568_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431568_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100002245.mm.1 | NR_045853.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100002245.mm.1 | NR_045853.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100002245.mm.1 | TC1100000261.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100002244.mm.1 | NR_045853.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100002244.mm.1 | NR_045853.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100002244.mm.1 | TC1100000261.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431568_at | - | - |
| Mouse Expression Set 430 | 1431568_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431568_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431568_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 309487 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 104120551 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2666653 | - | - |
| mouse-6 v1.1 (Illumina) | scl41762.9.1_46-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104120551 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104120551 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2666652 | - | - |

4933427E13Rik
Genomic Map