Assay Details
| Gene Symbol: Lyzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026092.3 | NP_080368.1 | 4 - 5 | 522 | 101 | |
| GenBank mRNA | AK006241.1 | - | 4 - 5 | 523 | 101 | |
| AK006630.1 | - | 4 - 5 | 523 | 101 | ||
| AY572447.1 | - | 4 - 5 | 481 | 101 | ||
| BC048450.1 | - | 4 - 5 | 493 | 6774504 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31461138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31461136 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31461137 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152696 | NM_026092.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152696 | AK006241.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152696 | AK006630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152696 | AY572447.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152696 | BC048450.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152699 | NM_026092.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152699 | AK006630.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2152699 | BC048450.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 707479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521868 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172503 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165443 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129130 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152696 | NM_026092.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152696 | AK006241.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152696 | AK006630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152696 | AY572447.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152696 | BC048450.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152699 | NM_026092.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152699 | AK006630.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2152699 | BC048450.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172935 | NM_026092.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172935 | AK006241.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172935 | AK006630.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172935 | AY572447.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172935 | BC048450.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31461781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31461778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31461779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31461785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31461789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31461795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31461787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31461782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31461793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31461776 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129130 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08897799 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165443 | - | - |
| Whole Mouse Genome G4122A | A_51_P172935 | NM_026092.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172935 | AK006241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172935 | AK006630.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172935 | AY572447.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172935 | BC048450.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132154 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129130 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165443 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08897799 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13844878 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13844876 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13948135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31461138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31461137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31461136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31461840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461855 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13851425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13851429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13809528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13923713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13809532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13851430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461914 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13887062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13992078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13809522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13809511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13809540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13844876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13851420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13809534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14093681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31461933 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10453614 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453612 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453613 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453615 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453610 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10453611 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449892_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10453609 | - | - |
| Mouse Genome 430 2.0 Array | 1449892_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449892_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67328_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67328_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800000073.mm.1 | NM_026092.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800000073.mm.1 | NM_026092.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800000073.mm.1 | TC1800000011.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449892_PM_at | - | - |
| Mouse Expression Set 430 | 1449892_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449892_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449892_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449892_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449892_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 319819 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6550324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6550324 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006550324 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2618942 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2618942 | - | - |
| mouse-6 v1.1 (Illumina) | scl52301.4.1_32-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6550324 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52301.4.1_32-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550324 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2618942 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2618942 | - | - |

Lyzl1
Genomic Map