Assay Details
| Gene Symbol: Gm4956 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002858.1 | 4 - 5 | 528 | 75 | ||
| GenBank mRNA | BC061226.1 | - | 4 - 5 | 528 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
glutathione S-transferase, alpha 3 |
Gsta3 | Gst2-3 | -22.6kb | Anti-sense | Chr. 1 - 21240585 - 21265575 on NCBI Build GRCm38 |
| Downstream | KH domain containing 1A | Khdc1a |
AU014713, C88118, Ndg1 |
38.1kb | Anti-sense | Chr. 1 - 21329476 - 21352200 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm4956
Genomic Map