Assay Details
| Gene Symbol: Tmem14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290679.1 | NP_001277608.1 | 3 - 4 | 505 | 85 | |
| NM_029398.3 | NP_083674.2 | 3 - 4 | 505 | 85 | ||
| GenBank mRNA | AK017734.1 | - | 3 - 4 | 420 | 85 | |
| BC034768.1 | - | 3 - 4 | 451 | 4039224 | 85 | |
| BC060034.1 | - | 3 - 4 | 449 | 6491675 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422893 | BC060034.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422893 | NM_029398.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422893 | AK017734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422893 | BC060034.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422893 | NM_029398.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422893 | AK017734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994733 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541743 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35827 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 553107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844096 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104667 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166685 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142086 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994733 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422893 | BC060034.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422893 | NM_029398.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422893 | AK017734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422893 | BC060034.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422893 | NM_029398.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422893 | AK017734.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20076755 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166685 | - | - |
| Whole Mouse Genome G4122A | A_51_P422893 | BC060034.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422893 | NM_029398.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422893 | AK017734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422893 | BC060034.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422893 | NM_029398.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422893 | AK017734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P339154 | BC034768.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P339154 | BC060034.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P339154 | NM_029398.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P339154 | AK017734.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124304 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166685 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01002548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01002540 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01002553 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01002535 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01002532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20076752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00002431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00206195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01073572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01002540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01002541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01002532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01002547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00621554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00175214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01002544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01002539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20076789 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10345059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345060 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428447_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10345058 | - | - |
| Mouse Genome 430 2.0 Array | 1428447_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428447_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75712_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75712_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100000935.mm.1 | NM_029398.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100000935.mm.1 | TC0100000133.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100000933.mm.1 | NM_029398.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100000933.mm.1 | TC0100000133.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428447_at | - | - |
| Mouse Expression Set 430 | 1428447_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428447_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428447_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6940301 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006940301 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2471444 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2471444 | - | - |
| mouse-6 v1.1 (Illumina) | scl18125.3.1_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6940301 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18125.3.1_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940301 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2471444 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2471444 | - | - |

Tmem14a
Genomic Map