Assay Details
| Gene Symbol: Tmem14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290679.1 | NP_001277608.1 | 4 - 5 | 587 | 67 | |
| GenBank mRNA | AK017734.1 | - | 4 - 5 | 502 | 67 | |
More Information
Set Membership: |
|

Tmem14a
Genomic Map