Assay Details
| Gene Symbol: Adamts7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003911.2 | NP_001003911.2 | 12 - 13 | 2102 | 94 | |
| GenBank mRNA | BC076575.1 | - | 11 - 12 | 1810 | 94 | |
| BC141173.1 | - | 12 - 13 | 1878 | 9055785 | 94 | |
| BC145491.1 | - | 12 - 13 | 1878 | 9054054 | 94 | |
| CB246893.1 | - | 3 - 4 | 95 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Adamts7
Genomic Map