Assay Details
| Gene Symbol: Adamts7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003911.2 | NP_001003911.2 | 13 - 14 | 2236 | 68 | |
| GenBank mRNA | AY551090.1 | - | 13 - 14 | 1969 | 68 | |
| BC058991.1 | - | 10 - 11 | 1214 | 68 | ||
| BC076575.1 | - | 12 - 13 | 1944 | 68 | ||
| BC141173.1 | - | 13 - 14 | 2012 | 9055785 | 68 | |
| BC145491.1 | - | 13 - 14 | 2012 | 9054054 | 68 | |
| CB246893.1 | - | 4 - 5 | 495 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adamts7
Genomic Map