Assay Details
| Gene Symbol: Adamts7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003911.2 | NP_001003911.2 | 17 - 18 | 2858 | 81 | |
| GenBank mRNA | AY551090.1 | - | 17 - 18 | 2591 | 81 | |
| BC058991.1 | - | 14 - 15 | 1830 | 81 | ||
| BC076575.1 | - | 16 - 17 | 2566 | 81 | ||
| BC141173.1 | - | 17 - 18 | 2634 | 9055785 | 81 | |
| BC145491.1 | - | 17 - 18 | 2634 | 9054054 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamts7
Genomic Map