| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900009114.mm.1 |
NM_001003911.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009114.mm.1 |
NM_001003911.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009114.mm.1 |
TC0900001158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900009120.mm.1 |
NM_001003911.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009120.mm.1 |
NM_001003911.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009120.mm.1 |
TC0900001158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900009115.mm.1 |
NM_001003911.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009115.mm.1 |
NM_001003911.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009115.mm.1 |
TC0900001158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900009116.mm.1 |
NM_001003911.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009116.mm.1 |
NM_001003911.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009116.mm.1 |
TC0900001158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900009122.mm.1 |
NM_001003911.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009122.mm.1 |
NM_001003911.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009122.mm.1 |
TC0900001158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900009118.mm.1 |
NM_001003911.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009118.mm.1 |
NM_001003911.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009118.mm.1 |
TC0900001158.mm.1 |
MTA TC |