Assay Details
| Gene Symbol: Adamts7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003911.2 | NP_001003911.2 | 4 - 5 | 1040 | 91 | |
| GenBank mRNA | AY551090.1 | - | 4 - 5 | 773 | 91 | |
| BC076575.1 | - | 4 - 5 | 894 | 91 | ||
| BC141173.1 | - | 4 - 5 | 816 | 9055785 | 91 | |
| BC145491.1 | - | 4 - 5 | 816 | 9054054 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamts7
Genomic Map