Assay Details
| Gene Symbol: Adamts7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003911.2 | NP_001003911.2 | 6 - 7 | 1250 | 70 | |
| GenBank mRNA | AY551090.1 | - | 6 - 7 | 983 | 70 | |
| BC030442.1 | - | 1 - 2 | 111 | 70 | ||
| BC058991.1 | - | 3 - 4 | 228 | 70 | ||
| BC076575.1 | - | 6 - 7 | 1104 | 70 | ||
| BC141173.1 | - | 6 - 7 | 1026 | 9055785 | 70 | |
| BC145491.1 | - | 6 - 7 | 1026 | 9054054 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamts7
Genomic Map