Assay Details
| Gene Symbol: Adamts7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003911.2 | NP_001003911.2 | 8 - 9 | 1546 | 82 | |
| GenBank mRNA | AY551090.1 | - | 8 - 9 | 1279 | 82 | |
| BC030442.1 | - | 3 - 4 | 407 | 82 | ||
| BC058991.1 | - | 5 - 6 | 524 | 82 | ||
| BC076575.1 | - | 8 - 9 | 1400 | 82 | ||
| BC141173.1 | - | 8 - 9 | 1322 | 9055785 | 82 | |
| BC145491.1 | - | 8 - 9 | 1322 | 9054054 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamts7
Genomic Map