Assay Details
| Gene Symbol: 4930524O08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040735.1 | 4 - 5 | 1154 | 75 | ||
| GenBank mRNA | AK015891.1 | - | 4 - 5 | 1155 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 184 | Mir184 |
Mirn184, mir-184, mmu-mir-184 |
-54.1kb | Anti-sense | Chr. 9 - 89802260 - 89802328 on NCBI Build GRCm38 |
| Downstream | RAS protein-specific guanine nucleotide-releasing factor 1 | Rasgrf1 |
AI844718, CDC25, CDC25Mm, Gnrp, Grf1, Grfbeta, P190-A, Ras-GRF1, p190, p190RhoGEF |
51.0kb | Sense | Chr. 9 - 89909472 - 90026979 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930524O08Rik
Genomic Map