Assay Details
| Gene Symbol: Ctsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007801.2 | NP_031827.2 | 3 - 4 | 510 | 98 | |
| GenBank mRNA | AK017638.1 | - | 3 - 4 | 242 | 98 | |
| AK149949.1 | - | 3 - 4 | 231 | 98 | ||
| AK150583.1 | - | 3 - 4 | 239 | 98 | ||
| AK151877.1 | - | 3 - 4 | 251 | 98 | ||
| AK157376.1 | - | 3 - 4 | 284 | 98 | ||
| AK160026.1 | - | 3 - 4 | 241 | 98 | ||
| AK171550.1 | - | 3 - 4 | 272 | 98 | ||
| BC006878.1 | - | 3 - 4 | 250 | 2647229 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctsh
Genomic Map