Assay Details
| Gene Symbol: Ctsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007801.2 | NP_031827.2 | 7 - 8 | 825 | 85 | |
| GenBank mRNA | AK017638.1 | - | 7 - 8 | 557 | 85 | |
| AK149949.1 | - | 7 - 8 | 546 | 85 | ||
| AK150583.1 | - | 7 - 8 | 554 | 85 | ||
| AK151877.1 | - | 7 - 8 | 566 | 85 | ||
| AK157376.1 | - | 7 - 8 | 599 | 85 | ||
| AK160026.1 | - | 7 - 8 | 556 | 85 | ||
| AK171550.1 | - | 7 - 8 | 587 | 85 | ||
| BC006878.1 | - | 7 - 8 | 565 | 2647229 | 85 | |
| U06119.1 | - | 7 - 8 | 589 | 85 | ||
| Y18464.1 | - | 2 - 3 | 137 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsh
Genomic Map