Assay Details
| Gene Symbol: Ctsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007801.2 | NP_031827.2 | 8 - 9 | 908 | 118 | |
| GenBank mRNA | AK017638.1 | - | 8 - 9 | 640 | 118 | |
| AK149949.1 | - | 8 - 9 | 629 | 118 | ||
| AK150583.1 | - | 8 - 9 | 637 | 118 | ||
| AK151877.1 | - | 8 - 9 | 649 | 118 | ||
| AK157376.1 | - | 8 - 9 | 682 | 118 | ||
| AK160026.1 | - | 8 - 9 | 639 | 118 | ||
| AK167498.1 | - | 7 - 8 | 533 | 118 | ||
| BC006878.1 | - | 8 - 9 | 648 | 2647229 | 118 | |
| U06119.1 | - | 8 - 9 | 672 | 118 | ||
| Y18464.1 | - | 3 - 4 | 220 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsh
Genomic Map