Assay Details
| Gene Symbol: Top3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009410.2 | NP_033436.1 | 11 - 12 | 1546 | 59 | |
| GenBank mRNA | AB006074.1 | - | 11 - 12 | 1547 | 59 | |
| AK029987.1 | - | 8 - 9 | 939 | 59 | ||
| AK037640.1 | - | 11 - 12 | 1447 | 59 | ||
| AK164443.1 | - | 11 - 12 | 1447 | 59 | ||
| BC099689.1 | - | 11 - 12 | 1361 | 30731611 | 59 | |
More Information
Set Membership: |
|

Top3a
Genomic Map