Assay Details
| Gene Symbol: Top3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009410.2 | NP_033436.1 | 12 - 13 | 1738 | 83 | |
| GenBank mRNA | AB006074.1 | - | 12 - 13 | 1739 | 83 | |
| AK029987.1 | - | 9 - 10 | 1131 | 83 | ||
| AK037640.1 | - | 12 - 13 | 1639 | 83 | ||
| AK164443.1 | - | 12 - 13 | 1639 | 83 | ||
| AK170291.1 | - | 13 - 14 | 1545 | 83 | ||
| AK171286.1 | - | 13 - 14 | 1545 | 83 | ||
| BC099689.1 | - | 12 - 13 | 1553 | 30731611 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top3a
Genomic Map