Assay Details
| Gene Symbol: Dpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133803.2 | NP_598564.2 | 4 - 5 | 571 | 71 | |
| GenBank mRNA | AK031020.1 | - | 4 - 5 | 536 | 71 | |
| AK039975.1 | - | 4 - 5 | 532 | 71 | ||
| AK087994.1 | - | 4 - 5 | 535 | 71 | ||
| AK149853.1 | - | 4 - 5 | 536 | 71 | ||
| AK150101.1 | - | 4 - 5 | 531 | 71 | ||
| AK150126.1 | - | 4 - 5 | 525 | 71 | ||
| AK166842.1 | - | 4 - 5 | 536 | 71 | ||
| AK168195.1 | - | 4 - 5 | 524 | 71 | ||
| BC004600.1 | - | 3 - 4 | 513 | 3584924 | 71 | |
| BC066817.1 | - | 4 - 5 | 522 | 30429618 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpp3
Genomic Map