Assay Details
| Gene Symbol: Dpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133803.2 | NP_598564.2 | 5 - 6 | 646 | 61 | |
| GenBank mRNA | AK031020.1 | - | 5 - 6 | 611 | 61 | |
| AK039975.1 | - | 5 - 6 | 607 | 61 | ||
| AK087994.1 | - | 5 - 6 | 610 | 61 | ||
| AK149853.1 | - | 5 - 6 | 611 | 61 | ||
| AK150101.1 | - | 5 - 6 | 606 | 61 | ||
| AK150126.1 | - | 5 - 6 | 600 | 61 | ||
| AK166842.1 | - | 5 - 6 | 611 | 61 | ||
| AK168195.1 | - | 5 - 6 | 599 | 61 | ||
| BC004600.1 | - | 4 - 5 | 588 | 3584924 | 61 | |
| BC066817.1 | - | 5 - 6 | 597 | 30429618 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpp3
Genomic Map