Assay Details
| Gene Symbol: Syt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134164.5 | NP_598925.1 | 1 - 2 | 212 | 94 | |
| GenBank mRNA | AK046627.1 | - | 1 - 2 | 516 | 94 | |
| AK083139.1 | - | 1 - 2 | 213 | 94 | ||
| AK140575.1 | - | 1 - 2 | 226 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046627.1, AK083139.1, NM_134164.5 transcripts.
Set Membership: |
|

Syt12
Genomic Map