Assay Details
| Gene Symbol: Syt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134164.5 | NP_598925.1 | 4 - 5 | 850 | 104 | |
| GenBank mRNA | AB062804.1 | - | 3 - 4 | 618 | 104 | |
| AK046627.1 | - | 4 - 5 | 1154 | 104 | ||
| AK083139.1 | - | 4 - 5 | 851 | 104 | ||
| BC116846.1 | - | 3 - 4 | 648 | 40126165 | 104 | |
| BC116848.1 | - | 3 - 4 | 648 | 40126167 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syt12
Genomic Map