Assay Details
| Gene Symbol: Syt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134164.5 | NP_598925.1 | 5 - 6 | 1068 | 71 | |
| GenBank mRNA | AB062804.1 | - | 4 - 5 | 836 | 71 | |
| AK046627.1 | - | 5 - 6 | 1372 | 71 | ||
| AK083139.1 | - | 5 - 6 | 1069 | 71 | ||
| BC116846.1 | - | 4 - 5 | 866 | 40126165 | 71 | |
| BC116848.1 | - | 4 - 5 | 40126167 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syt12
Genomic Map