Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 11 - 12 | 1514 | 80 | |
| NM_008797.3 | NP_032823.2 | 10 - 11 | 1491 | 80 | ||
| GenBank mRNA | AK077665.1 | - | 2 - 3 | 319 | 80 | |
| AK128927.1 | - | 1441 | 80 | |||
| AK148328.1 | - | 11 - 12 | 1461 | 80 | ||
| AK170596.1 | - | 11 - 12 | 1474 | 80 | ||
| AK170871.1 | - | 11 - 12 | 1464 | 80 | ||
| AK172320.1 | - | 11 - 12 | 1442 | 80 | ||
| BC055030.1 | - | 11 - 12 | 1449 | 5053475 | 80 | |
| L09192.1 | - | 10 - 11 | 1467 | 80 | ||
| M97957.1 | - | 5 - 6 | 639 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map