Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 12 - 13 | 1657 | 59 | |
| NM_008797.3 | NP_032823.2 | 11 - 12 | 1634 | 59 | ||
| GenBank mRNA | AK077665.1 | - | 3 - 4 | 462 | 59 | |
| AK128927.1 | - | 1584 | 59 | |||
| AK148328.1 | - | 12 - 13 | 1604 | 59 | ||
| AK170596.1 | - | 12 - 13 | 1617 | 59 | ||
| AK170871.1 | - | 12 - 13 | 1607 | 59 | ||
| AK172320.1 | - | 12 - 13 | 1585 | 59 | ||
| AK182295.1 | - | 1 - 2 | 25 | 59 | ||
| BC055030.1 | - | 12 - 13 | 1592 | 5053475 | 59 | |
| L09192.1 | - | 11 - 12 | 1610 | 59 | ||
| M97957.1 | - | 6 - 7 | 782 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map