Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 14 - 15 | 2125 | 75 | |
| NM_008797.3 | NP_032823.2 | 13 - 14 | 2102 | 75 | ||
| GenBank mRNA | AK077665.1 | - | 5 - 6 | 930 | 75 | |
| AK128927.1 | - | 2052 | 75 | |||
| AK148328.1 | - | 14 - 15 | 2072 | 75 | ||
| AK170596.1 | - | 14 - 15 | 2085 | 75 | ||
| AK170871.1 | - | 14 - 15 | 2075 | 75 | ||
| AK172320.1 | - | 14 - 15 | 2053 | 75 | ||
| BC055030.1 | - | 14 - 15 | 2060 | 5053475 | 75 | |
| L09192.1 | - | 13 - 14 | 2078 | 75 | ||
| M97957.1 | - | 8 - 9 | 1250 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map