Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 16 - 17 | 2618 | 89 | |
| NM_008797.3 | NP_032823.2 | 15 - 16 | 2595 | 89 | ||
| GenBank mRNA | AK077665.1 | - | 7 - 8 | 1423 | 89 | |
| AK148328.1 | - | 16 - 17 | 2565 | 89 | ||
| AK170596.1 | - | 16 - 17 | 2578 | 89 | ||
| AK170871.1 | - | 16 - 17 | 90 | |||
| AK172320.1 | - | 16 - 17 | 2546 | 89 | ||
| BC055030.1 | - | 16 - 17 | 2553 | 5053475 | 89 | |
| L09192.1 | - | 15 - 16 | 2571 | 89 | ||
| M97957.1 | - | 10 - 11 | 1743 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map