Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 17 - 18 | 2871 | 93 | |
| NM_008797.3 | NP_032823.2 | 16 - 17 | 2848 | 93 | ||
| GenBank mRNA | AK077665.1 | - | 8 - 9 | 1676 | 93 | |
| AK128927.1 | - | 2645 | 93 | |||
| AK148328.1 | - | 17 - 18 | 2818 | 93 | ||
| AK170596.1 | - | 17 - 18 | 2831 | 93 | ||
| AK170871.1 | - | 17 - 18 | 2822 | 93 | ||
| AK172320.1 | - | 17 - 18 | 2799 | 93 | ||
| BC055030.1 | - | 17 - 18 | 2806 | 5053475 | 93 | |
| L09192.1 | - | 16 - 17 | 2824 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map