Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 18 - 19 | 3048 | 62 | |
| NM_008797.3 | NP_032823.2 | 17 - 18 | 3025 | 62 | ||
| GenBank mRNA | AK077665.1 | - | 9 - 10 | 1853 | 62 | |
| AK128927.1 | - | 2822 | 62 | |||
| AK148328.1 | - | 18 - 19 | 2995 | 62 | ||
| AK170596.1 | - | 18 - 19 | 3008 | 62 | ||
| AK170871.1 | - | 18 - 19 | 2999 | 62 | ||
| AK172320.1 | - | 18 - 19 | 2976 | 62 | ||
| BC055030.1 | - | 18 - 19 | 2983 | 5053475 | 62 | |
| L09192.1 | - | 17 - 18 | 3001 | 62 | ||
| M97957.1 | - | 12 - 13 | 2173 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map