Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008797.3 | NP_032823.2 | 1 - 2 | 131 | 150 | |
| GenBank mRNA | BY139237.1 | - | 1 - 2 | 129 | 150 | |
| L09192.1 | - | 1 - 2 | 107 | 150 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the L09192.1, NM_008797.3 transcripts.
Set Membership: |
|

Pcx
Genomic Map