Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 5 - 6 | 638 | 74 | |
| NM_008797.3 | NP_032823.2 | 4 - 5 | 615 | 74 | ||
| GenBank mRNA | AK128927.1 | - | 565 | 74 | ||
| AK148328.1 | - | 5 - 6 | 585 | 74 | ||
| AK170596.1 | - | 5 - 6 | 598 | 74 | ||
| AK170871.1 | - | 5 - 6 | 588 | 74 | ||
| AK172320.1 | - | 5 - 6 | 566 | 74 | ||
| BC055030.1 | - | 5 - 6 | 573 | 5053475 | 74 | |
| BG087228.2 | - | 5 - 6 | 574 | 74 | ||
| L09192.1 | - | 4 - 5 | 591 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map