Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 7 - 8 | 895 | 63 | |
| NM_008797.3 | NP_032823.2 | 6 - 7 | 872 | 63 | ||
| GenBank mRNA | AK128927.1 | - | 822 | 63 | ||
| AK148328.1 | - | 7 - 8 | 842 | 63 | ||
| AK170596.1 | - | 7 - 8 | 855 | 63 | ||
| AK170871.1 | - | 7 - 8 | 845 | 63 | ||
| AK172320.1 | - | 7 - 8 | 823 | 63 | ||
| BC055030.1 | - | 7 - 8 | 830 | 5053475 | 63 | |
| L09192.1 | - | 6 - 7 | 848 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map