Assay Details
| Gene Symbol: Rce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023131.1 | NP_075620.1 | 2 - 3 | 291 | 64 | |
| GenBank mRNA | AJ251645.1 | - | 2 - 3 | 291 | 64 | |
| AK012946.1 | - | 2 - 3 | 247 | 64 | ||
| BC048686.1 | - | 2 - 3 | 315 | 64 | ||
More Information
Set Membership: |
|

Rce1
Genomic Map