Assay Details
| Gene Symbol: Klc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008451.2 | NP_032477.2 | 14 - 15 | 2023 | 107 | |
| NR_045528.1 | 14 - 15 | 2055 | 107 | |||
| GenBank mRNA | AF055666.1 | - | 14 - 15 | 1847 | 104 | |
| AK141439.1 | - | 14 - 15 | 1948 | 107 | ||
| BC014845.1 | - | 14 - 15 | 1867 | 2599082 | 107 | |
| CX565742.1 | - | 6 - 7 | 623 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klc2
Genomic Map