Assay Details
| Gene Symbol: Kdm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001984.2 | NP_001001984.2 | 3 - 4 | 1005 | 90 | |
| GenBank mRNA | AK037157.1 | - | 4 - 5 | 902 | 90 | |
| AK147028.1 | - | 3 - 4 | 997 | 90 | ||
| AK155866.1 | - | 3 - 4 | 1010 | 90 | ||
| BC076576.1 | - | 3 - 4 | 571 | 6840591 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm2a
Genomic Map