Assay Details
| Gene Symbol: Kdm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001984.2 | NP_001001984.2 | 6 - 7 | 1316 | 74 | |
| GenBank mRNA | AK037157.1 | - | 7 - 8 | 1213 | 74 | |
| AK147028.1 | - | 6 - 7 | 1308 | 74 | ||
| AK155866.1 | - | 6 - 7 | 1321 | 74 | ||
| BC076576.1 | - | 6 - 7 | 882 | 6840591 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm2a
Genomic Map