Assay Details
| Gene Symbol: Kdm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001984.2 | NP_001001984.2 | 7 - 8 | 1413 | 110 | |
| GenBank mRNA | AK037157.1 | - | 8 - 9 | 1310 | 110 | |
| AK147028.1 | - | 7 - 8 | 1405 | 110 | ||
| BC076576.1 | - | 7 - 8 | 979 | 6840591 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm2a
Genomic Map