Assay Details
| Gene Symbol: Adrbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290818.1 | NP_001277747.1 | 9 - 10 | 953 | 78 | |
| NM_130863.2 | NP_570933.1 | 10 - 11 | 1025 | 78 | ||
| GenBank mRNA | AF333028.1 | - | 9 - 10 | 745 | 78 | |
| AK155691.1 | - | 9 - 10 | 954 | 78 | ||
| BC003196.1 | - | 9 - 10 | 741 | 78 | ||
| BC033272.1 | - | 8 - 9 | 1069 | 78 | ||
| BC053922.1 | - | 10 - 11 | 758 | 6309555 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adrbk1
Genomic Map