Assay Details
| Gene Symbol: Ssh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198113.2 | NP_932781.1 | 12 - 13 | 1491 | 71 | |
| GenBank mRNA | AB099289.1 | - | 12 - 13 | 1489 | 71 | |
| AK038981.1 | - | 10 - 11 | 2705 | 71 | ||
| AK148293.1 | - | 12 - 13 | 1484 | 71 | ||
| AK149880.1 | - | 12 - 13 | 1512 | 71 | ||
| AK164662.1 | - | 11 - 12 | 1190 | 71 | ||
| AK166342.1 | - | 12 - 13 | 1498 | 71 | ||
| BC028922.1 | - | 12 - 13 | 1491 | 3987714 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ssh3
Genomic Map