Assay Details
| Gene Symbol: Ssh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198113.2 | NP_932781.1 | 4 - 5 | 549 | 130 | |
| GenBank mRNA | AB099289.1 | - | 4 - 5 | 547 | 130 | |
| AK038981.1 | - | 3 - 4 | 1121 | 130 | ||
| AK148293.1 | - | 4 - 5 | 542 | 130 | ||
| AK149880.1 | - | 4 - 5 | 558 | 130 | ||
| AK164662.1 | - | 3 - 4 | 247 | 130 | ||
| AK166342.1 | - | 4 - 5 | 555 | 130 | ||
| BC028922.1 | - | 4 - 5 | 549 | 3987714 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ssh3
Genomic Map