Assay Details
| Gene Symbol: Ssh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198113.2 | NP_932781.1 | 8 - 9 | 974 | 104 | |
| GenBank mRNA | AB099289.1 | - | 8 - 9 | 972 | 104 | |
| AK038981.1 | - | 7 - 8 | 1546 | 104 | ||
| AK148293.1 | - | 8 - 9 | 967 | 104 | ||
| AK149880.1 | - | 8 - 9 | 995 | 104 | ||
| AK164662.1 | - | 7 - 8 | 672 | 104 | ||
| AK166342.1 | - | 8 - 9 | 981 | 104 | ||
| BC028922.1 | - | 8 - 9 | 974 | 3987714 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ssh3
Genomic Map