Assay Details
| Gene Symbol: Cnih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB006191.2 | - | 3 - 4 | 351 | 87 | |
| AK141483.1 | - | 3 - 4 | 451 | 87 | ||
| BC059922.1 | - | 3 - 4 | 437 | 87 | ||
| BC103797.1 | - | 3 - 4 | 425 | 30134114 | 87 | |
| BC137585.1 | - | 3 - 4 | 312 | 8860604 | 87 | |
| BC137586.1 | - | 3 - 4 | 312 | 8860605 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnih2
Genomic Map