Assay Details
| Gene Symbol: Il1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010555.4 | NP_034685.1 | 5 - 6 | 812 | 114 | |
| GenBank mRNA | AK132264.1 | - | 5 - 6 | 839 | 114 | |
| BC032962.1 | - | 4 - 5 | 529 | 1282531 | 114 | |
| CT010182.1 | - | 4 - 5 | 720 | 114 | ||
| X59769.1 | - | 5 - 6 | 812 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il1r2
Genomic Map