Assay Details
| Gene Symbol: Il1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010555.4 | NP_034685.1 | 7 - 8 | 1009 | 74 | |
| GenBank mRNA | AK132264.1 | - | 7 - 8 | 1036 | 74 | |
| CT010182.1 | - | 6 - 7 | 917 | 74 | ||
| X59769.1 | - | 7 - 8 | 1009 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il1r2
Genomic Map