Assay Details
| Gene Symbol: Npas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008719.2 | NP_032745.2 | 3 - 4 | 680 | 77 | |
| GenBank mRNA | BC109166.2 | - | 3 - 4 | 680 | 40048145 | 77 |
| U77969.1 | - | 3 - 4 | 754 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npas2
Genomic Map