Assay Details
| Gene Symbol: Uprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081189.1 | NP_001074658.1 | 6 - 7 | 1010 | 122 | |
| GenBank mRNA | BC147845.1 | - | 6 - 7 | 868 | 9087562 | 122 |
| BC147853.1 | - | 6 - 7 | 868 | 9087570 | 122 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965821 | BC147853.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965821 | NM_001081189.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965821 | BC147845.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 130035 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499586 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10688 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722739 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491343 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25429 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916014 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381740 | AK139666.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381740 | BC023868.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381740 | BC058590.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381740 | BC099495.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381740 | NM_001114102.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 170757 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40211 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38176 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965821 | BC147853.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965821 | NM_001081189.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965821 | BC147845.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32577528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15220250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15117065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15008598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14521442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14661201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14526195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577514 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14969744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14526190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15080937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15145764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14630753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577496 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15193858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14521439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14521447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15145762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14580947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14930227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14521438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14761609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14835032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14930228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14769284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14630757 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32577503 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601333 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601334 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601329 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601332 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601326 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601328 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 500521 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2936221 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2936221 | - | - |

Uprt
Genomic Map