Assay Details
| Gene Symbol: Tmem182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081198.1 | NP_001074667.1 | 2 - 3 | 389 | 77 | |
| GenBank mRNA | BC147440.1 | - | 2 - 3 | 346 | 9056082 | 77 |
| BC147458.1 | - | 2 - 3 | 346 | 9056100 | 77 | |
| BC147740.1 | - | 2 - 3 | 346 | 9056689 | 77 | |
| BC147743.1 | - | 2 - 3 | 346 | 9056692 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem182
Genomic Map